Bis(2-ethoxyethyl)ether 99+% for HPLC

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

43464.AK
43464.AKEA 136 GBP
43464.AK
Bis(2-ethoxyethyl)ether 99+% for HPLC
Bis(2-ethoxyethyl) ether

Purity: 99+% CAS No.: 112-36-7 Molecular Formula: (CH3CH2OCH2CH2)2O Formula Weight: 162.23 Flash Point: 71°(160°F) Physical Form: Liquid Density: 0.909 Refractive Index: 1.4120 Melting Point: -44° Boiling Point: 188-190° Beilstein Reference: 1699259 Merck Reference: 14,3118 MDL No.: MFCD00009254

Formula: C₈H₁₈O₃
Boiling Pt: 188…190 °C
Melting Pt: –44 °C
Density: 0.909
Flash Pt: 71 °C (160 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009254
CAS Number: 112-36-7
EINECS: 203-963-7
Merck Index: 14,3118

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR