2-Thienylmagnesium bromide 1 M in THF, AcroSeal®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: 2-Thienylmagnesium bromide

434610500
434610500.EA 82.2 GBP
434610500.
2-Thienylmagnesium bromide 1 M in THF, AcroSeal®
2-Thienylmagnesium bromide
Formula: C₄H₃BrMgS
MW: 187.34 g/mol
Boiling Pt: 65 °C (760 mmHg)
Density: 1.011 g/cm³
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD03094046
CAS Number: 5713-61-1
UN: 2924
ADR: 3 II

Order Now

Specification Test Results

Appearance Yellow to orange to brown Solution
Titration Grignard 0.95 to 1.10 M
Melting point 34 to 38 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR