Ruthenium(III) nitrosylsulphate ≥99.99% (metals basis), Premion®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

43436.03 43436.06
43436.03EA 81.4 GBP
43436.03 43436.06
Ruthenium(III) nitrosylsulphate ≥99.99% (metals basis), Premion®
Ruthenium(III) nitrosylsulphate

Purity: 99.99% Molecular Formula: [Ru(NO)]2(SO4)3 Formula Weight: 550.34 Physical Form: Powder Solubility: Soluble in water MDL No.: MFCD02684508


Applications: Electroplating, coating

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR