Strontium iodide, anhydrous ≥99.99% (metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

43374.22 43374.14 43374.06
43374.22EA 1067 GBP
43374.22 43374.14 43374.06
Strontium iodide, anhydrous ≥99.99% (metals basis)
Strontium iodide

Purity: 99.99% CAS No.: 10476-86-5 Molecular Formula: SrI2 Formula Weight: 341.43 Physical Form: Powder Density: 4.550 Appearance: White to pale yellow Odour: Odourless Melting Point: 515° Merck Reference: 14,8844 MDL No.: MFCD00049557

Formula: I₂Sr
MW: 341.43 g/mol
Melting Pt: 515
Density: 4.550
Storage Temperature: Ambient
MDL Number: MFCD00049557
CAS Number: 10476-86-5
EINECS: 233-972-1
UN: 3260
ADR: 8 II
Merck Index: 14,8844

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR