Paraformaldehyde 16% (w/v) in aqueous solution methanol-free

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

43368.9L 43368.9M
43368.9LEA 41.4 GBP
43368.9L 43368.9M
Paraformaldehyde 16% (w/v) in aqueous solution methanol-free
Formaldehyde

Solution is micro-filtered and supplied in ampules. RNase and DNase free.


CAS No.: 30525-89-4 Molecular Formula: (CH2O)n Physical Form: Liquid, Ampouled under argon MDL No.: MFCD00133991

Formula: CH₂O
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 3334
ADR: 9,III

Order Now

Specification Test Results

Concentration (w/v) 15.5 to 16.5%
Appearance Clear colourless liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR