Palladium ≥99.95% (metals basis), granules

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

43353.03 43353.14 43353.06
43353.03EA 301 GBP
43353.03 43353.14 43353.06
Palladium ≥99.95% (metals basis), granules
Palladium

Purity: 99.95% CAS No.: 7440-05-3 Odour: Odourless MDL No.: MFCD00011167

Formula: Pd
Boiling Pt: 3167 °C (1013 hPa)
Melting Pt: 1555 °C
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
Merck Index: 12,07121

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR