Molybdenum ≥99.95% (metals basis), wire, annealed, Ø 1.6 mm (0.063 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

43311.G1 43311.G9 43311.G5
43311.G1EA 53.2 GBP
43311.G1 43311.G9 43311.G5
Molybdenum ≥99.95% (metals basis), wire, annealed, Ø 1.6 mm (0.063 in)
Molybdenum

Purity: 99.95% CAS No.: 7439-98-7 Physical Form: ≈20.5g/m, Temper: soft MDL No.: MFCD00003465

Formula: Mo
Boiling Pt: 3617 °C (1013 hPa)
Melting Pt: 2622 °C
Density: 10.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR