Tin ≥99.9% (metals basis), foil, Thickness: 0.18 mm (0.007 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

43308.HP 43308.GH 43308.FI
43308.HPEA 352 GBP
43308.HP 43308.GH 43308.FI
Tin ≥99.9% (metals basis), foil, Thickness: 0.18 mm (0.007 in)
Tin

Purity: 99.9% CAS No.: 7440-31-5 Physical Form: ≈3.3g/50x50mm Odour: Odourless MDL No.: MFCD00133862

Formula: Sn
Boiling Pt: 2270 °C (1013 hPa)
Melting Pt: 232 °C
Density: 7.31 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133862
CAS Number: 7440-31-5
EINECS: 231-141-8
Merck Index: 12,09587

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR