Grinding pieces, zirconium oxide, yttria stabilised

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

43286.A1 43285.A1 43286.30 43285.30
43286.A1EA 487 GBP
43286.A1 43285.A1 43286.30 43285.30
Grinding pieces, zirconium oxide, yttria stabilised
Beads Ceramic Beads
The density of the zirconium oxide beads is 6.02 g/cm³

  • MDL No.: MFCD00145546
  • Yttrium oxide (mole) 3%
  • Hardness 12
  • Total impuroty level < 0.1

Storage at ambient temperatures.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR