Zinc chloride 97,0-100,5, granular USP, Multi-Compendial, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride

4326-01EA 228 GBP
4326-01 4326-05
Zinc chloride 97,0-100,5, granular USP, Multi-Compendial, J.T.Baker®
Zinc chloride

  • GMP manufactured product

Formula: ZnCl₂
MW: 136.3 g/mol
Boiling Pt: 732
Melting Pt: 283
Density: 2.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8 III
Merck Index: 13,10185

Order Now

Specification Test Results

GMP Manufactured Product Passes Test
Meets U.S.P Requirements Passes Test
Meets E.P. Chemical Specifications ≤0.03%
Meets J.P. Chemical Specifications ≤1.0%
Meets B.P. Chemical Specifications Passes Test
Store at 2 to 25 °C ≤0.005%
CAUTION: For Manufacturing, processing or repackaging 97.0 - 100.5%
Bulk Pharmaceutical Chemical ≤10
USP - Identification Passes Test
USP - Oxychloride Passes Test
USP - Sulfate (SO₄) ≤0.03 %
USP - Alkalies and Alkaline Earths ≤1.0 %
USP - Ammonium Salts Passes Test
USP - Lead (Pb) ≤0.005 %
USP - Assay (ZnCl₂) 97.0 - 100.5 %
Endotoxin Concentration (EU/g) ≤10
EP/BP - Assay (ZnCl₂) 95.0 - 100.5 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - pH 4.6 - 5.5
EP/BP - Oxychloride Passes Test
EP/BP - Sulfate (SO₄) ≤200 ppm
EP/BP - Aluminum, Calcium, Heavy Metals, Iron and Magnesium Passes Test
EP/BP - Ammonium (NH₄) ≤400 ppm
JP - Assay (ZnCl₂) 97.0 - 101.0 %
JP - Identification Passes Test
JP - Clarity and Color of Solution Passes Test
JP - Sulfate (SO₄) ≤0.010 %
JP - Ammonium (NH₄) Passes Test
JP - Heavy Metals (as Pb) ≤50 ppm
JP - Alkalies and Alkaline Earths ≤1.00 %
JP - Arsenic (As) ≤5 ppm
JP - Oxychloride Passes Test
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR