Samarium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 1.0 mm (0.04 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

43217.FF 43217.FI
43217.FFEA 304 GBP
43217.FF 43217.FI
Samarium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 1.0 mm (0.04 in)
Samarium

Purity: 99.9% CAS No.: 7440-19-9 Physical Form: ≈4.9g/25x25mm, Packaged in mylar under argon Odour: Odourless MDL No.: MFCD00011233

Formula: Sm
Boiling Pt: 1900 °C (1013 hPa)
Melting Pt: 1076.85 °C
Density: 7.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011233
CAS Number: 7440-19-9
EINECS: 231-128-7
Merck Index: 12,08498

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR