Tissue grinders
Supplier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
196102 196105 196110 196115 196302 196305 196310 196315
432-1202EA 126 GBP
432-1202 432-1205 432-1210 432-1215 432-1235 432-1240 432-1245 432-1250
Tissue grinders
Homogenisers Tissue Grinders
Clearance between Glass vessel and appropriately sized plunger is approximately 200 µm.