Laboratory blenders, BagMixer® 400
Supplier: INTERSCIENCE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
021230 022230 023230 024230 025000 025100
INSC021230EA 2510 GBP
INSC021230 INSC022230 INSC023230 432-0313 432-0315 432-0314
Laboratory blenders, BagMixer® 400
Homogenisers Laboratory Paddle Blenders
BagMixer® are highly efficient laboratory blenders for solid sample preparation. With a sterile bag, they guarantee optimal bacterial extraction in less than 60 seconds and eliminate all risks of cross contamination.
  • Door opens at 270° and paddles stop side be side at end of blending for easy extraction of bags
  • Removable paddles and stainless steel construction for ease of cleaning
  • Easy control of blending time, power and speed with VW CC, S and SW models
  • Q-Tight®, a special closing system prevents risks of leaking liquid from the top of the bag on VW and CC models