Analytical mill, A 11 basic
Supplier: IKA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2900000 2900002
412-1111EA 2340 GBP
412-1111 432-0027
Analytical mill, A 11 basic
Mills
This batch mill performs two different types of grinding; impact grinding of hard, brittle or non-elastic materials by means of the high-grade stainless steel beater, which can be used for samples with a Mohs hardness of up to six. The A 11 also provides cutting grinding, ideal for pulverising soft, fibrous materials with an optional cutting blade.
  • Two different types of grinding; impact and cutting
  • Damp and gelatinous materials can be pulverised by adding water
  • ETFE and reinforced glass fibre grinding chamber can be used with liquid nitrogen (for
More Product Information