3,4-Dimethylphenylmagnesium bromide 0.5 M in THF, AcroSeal®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

431910500
431910500.EA 111 GBP
431910500.
3,4-Dimethylphenylmagnesium bromide 0.5 M in THF, AcroSeal®
3,4-Dimethylphenylmagnesium bromide
Formula: C₈H₉BrMg
Boiling Pt: 65 °C (760 mmHg)
Density: 0.91 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD01311496
CAS Number: 89980-68-7
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Yellow to brown Solution
Titration Grignard 0.40 to 0.60 M
Refractive index 1.4590 to 1.4610 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR