ICP-MS internal standard for 6020, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

43161.AEEA 154 GBP
43161.AE
ICP-MS internal standard for 6020, Specpure®
Standards Cationic Standards Multi-Element Standards

Molecular Formula: Matrix: 5% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD01868829

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR