Homogeniser, POLYTRON®, PT 2500 E
Supplier: KINEMATICA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
11010055 11010056 11010057
431-1268EA 1510 GBP
431-1268 431-1269 431-1270
Homogeniser, POLYTRON®, PT 2500 E
Homogenisers Homogenisers Bench
Stand model, volume range 0,05 to 2500 ml for dispersing, homogenising, extractions, cell disruption, mixing, emulsifying and suspending. This model replaces the PT 1600 E and the PT 2100 incorporating the best aspects of both models.
  • Quick release coupling system for the dispersing elements (type E)
  • Integrated electronic speed regulation/stabilisation with control knob and large LED speed indicator
  • Speed remains constant even when the viscosity changes
  • Soft start - progressive speed increase
Certifications: IP protection class according to DIN EN 6052More Product Information