Homogeniser T 25 Easy Clean Digital
Supplier: IKA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
25002560 25002562
431-0314EA 3180 GBP
431-0314 431-0315
Homogeniser T 25 Easy Clean Digital
Homogenisers Homogenisers Bench
The T 25 easy clean digital is a high-performance dispersing instrument with electronic speed control from 3000 - 25000 rpm for working volumes of 1 - 2000 ml (H2O).
  • No disassembly required
  • Brushless motor guarantees dust-free and silent operation
  • USB interface for control and documentation through labworldsoft®
  • Timer/counter function
  • Increased safety through tool recognition and IP-class
The S 25 dispersing elements allows the T 25 to be used in various applications- whether it is homogenizing medical samples, emulsifying cosmetics or the suspensionMore Product Information