Graphite ≥97% (metals basis), foil, Thickness: 1 mm (0.04 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

43083.CJ 43083.RF 43083.RX
43083.CJEA 80 GBP
43083.CJ 43083.RF 43083.RX
Graphite ≥97% (metals basis), foil, Thickness: 1 mm (0.04 in)
Graphite

Purity: 97% MDL No.: MFCD00144065

Formula: C
Melting Pt: 3827 °C
Density: 2.09…2.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR