J.T.Baker® Plate, SRW Magnetic
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
43001-0505EA 315 GBP
43001-0505 43001-0504 43001-0506 43001-0514 43001-0515 43001-0516
J.T.Baker® Plate, SRW Magnetic
Microplates HTS Microplates
These plates are manufactured from high purity PP for efficient sample preparation and are suitable for use with magnetic particle processors such as the KingFisher™ Apex, Flex 96 and Presto.
  • Standard SBS footprint
  • Cylindrical wells enable good sample mixing
  • Slightly raised rims reduce the risk of cross-contamination
  • Alphanumeric code assists registration and identification of individual samples