J.T.Baker®, PCR Tubes and Caps for qPCR, 8-Strips
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
43001-0033EA 103 GBP
43001-0033 43001-0034
J.T.Baker®, PCR Tubes and Caps for qPCR, 8-Strips
Tubes PCR Tubes
PCR 8-tube strips and 8-cap strips are made of high-quality PP resin. Clear caps are compatible with qPCR application.
  • Certified free from DNase, RNase and human DNA
  • White 8-tube strips
  • Corresponding 8-cap strips, clear to be ordered separately
  • Ideal for low volume PCR and qPCR applications
  • Low profile tubes help to reduce condensation
  • Validated on Biorad CFX device
PCR strips and caps are manufactured under clean room conditions according to DIN EN ISO 14644-1:2016-06.
Packaging: 120 pieces of tubes and caps.