Strontium ≥99.8% (metals basis), pieces, distilled dendritic

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger


Ampuled under argon

42929.06 42929.14
42929.06EA 411 GBP
42929.06 42929.14
Strontium ≥99.8% (metals basis), pieces, distilled dendritic
Strontium

Purity: 99.8% CAS No.: 7440-24-6 Physical Form: Ampouled under argon Sensitivity: Air and Moisture Sensitive Appearance: Gold UN No.: 3208 MDL No.: MFCD00134060

Formula: Sr
MW: 87.62 g/mol
Boiling Pt: 1151
Melting Pt: 570…660 °C
Density: 2.38 g/cm³ (770 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134060
CAS Number: 7440-24-6
EINECS: 231-133-4
UN: 3208
ADR: 4.3 II
Merck Index: 12,08994

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR