2,7,12,17-Tetra-tert-butyl-5,10,15,20-tetraaza-21H,23H-porphine, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

428361-250MG
428361-250MGEA 178 GBP
428361-250MG
2,7,12,17-Tetra-tert-butyl-5,10,15,20-tetraaza-21H,23H-porphine, Sigma-Aldrich®
2,7,12,17-Tetra-tert-butyl-5,10,15,20-tetraaza-21H,23H-porphine

2,7,12,17-Tetra-tert-butyl-5,10,15,20-tetraaza-21H,23H-porphine has been used in multifunctional drug delivery investigations from ionic triblock terpolymer micelles.

Formula: C₃₂H₄₂N₈
MW: 538.73 g/mol
Melting Pt: 182 °C (dec., lit.)
Storage Temperature: Ambient
MDL Number: MFCD00209614
CAS Number: 64987-70-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6bc3e65f03ebeb2f38f7d288889803d52c3a86e4&vid=333ede60fe794297b6370f95f0ddac93a353c52f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR