Boron trichloride 10% w/w 1 M 2-chloroethanol, AcroSeal®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

428331000
428331000.EA 49.7 GBP
428331000.
Boron trichloride 10% w/w 1 M 2-chloroethanol, AcroSeal®
Boron trichloride
Formula: BCl₃
MW: 117.17 g/mol
Boiling Pt: 12.4
Melting Pt: -107
Density: 1.26 g/cm³
Flash Pt: 53 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011313
CAS Number: 10294-34-5
UN: 3390
ADR: 6.1
Merck Index: 12,01378

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Titration Argentometric 0.9 to 1.1 M
Melting point 62 to 66 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR