Methyl purple 0.1% (w/v) in aqueous solution

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

42753.AP 42753.K2
42753.APEA 39.5 GBP
42753.AP 42753.K2
Methyl purple 0.1% (w/v) in aqueous solution
Methyl purple

Physical Form: Liquid MDL No.: MFCD00070843

Formula: C₆₇H₆₂N₈Na₄O₁₄S₃
Storage Temperature: Ambient
MDL Number: MFCD00070843
CAS Number: 1340-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR