Conductivity standards, KCl, referenced at 25 °C
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
42679.AP 42679.K2 42692.AP 42692.K2 42692.K7 42693.AP 42693.K2 42693.K7 42694.AP 42694.K2 42695.AP 42695.K2 42695.K7 42696.AP 42696.K2 42697.AP 42680.K2 43405.K2 42680.K7 43405.AP 42680.AP 47057.AP 47057.K2
42679.APEA 31.3 GBP
42679.AP 42679.K2 42692.AP 42692.K2 42692.K7 42693.AP 42693.K2 42693.K7 42694.AP 42694.K2 42695.AP 42695.K2 42695.K7 42696.AP 42696.K2 42697.AP 42680.K2 43405.K2 42680.K7 43405.AP 42680.AP 47057.AP 47057.K2
Conductivity standards, KCl, referenced at 25 °C
Standards Conductivity Standards
KCl conductivity standards used for calibrating conductivity cells with ranging from 718 to 100 000 µS/cm.