Buffer Solutions

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

38712.AP 38712.K2 38712.K7 38714.AP 38714.K2 38714.K7 40439.AP 40439.K2 40439.K7 40447.AP 40447.K2 40447.K7 42416.AP 42416.K2 42416.K7 42417.AP 42417.K2 42417.K7 42418.AP 42418.K2 42418.K7 42419.AP 42419.K2 42419.K7 42420.AP 42420.K2 42420.K7 42421.AP 42421.K2 42421.K7 42422.AP 42422.K2 42422.K7 42423.AP 42423.K2 42424.AP 42424.K2 42424.K7 42425.AP 42425.K2 42425.K7 46307.AP 46307.K2 46307.K7
38712.APEA 20.6 GBP
38712.AP 38712.K2 38712.K7 38714.AP 38714.K2 38714.K7 40439.AP 40439.K2 40439.K7 40447.AP 40447.K2 40447.K7 42416.AP 42416.K2 42416.K7 42417.AP 42417.K2 42417.K7 42418.AP 42418.K2 42418.K7 42419.AP 42419.K2 42419.K7 42420.AP 42420.K2 42420.K7 42421.AP 42421.K2 42421.K7 42422.AP 42422.K2 42422.K7 42423.AP 42423.K2 42424.AP 42424.K2 42424.K7 42425.AP 42425.K2 42425.K7 46307.AP 46307.K2 46307.K7
Buffer Solutions
Buffers pH Calibration Buffers

Order Now

SPECIFICATIONS

Environmentally Preferable
Odour/scent Odourless Odourless Odourless
Physical form Liquid Liquid Liquid Liquid Liquid Liquid Liquid Liquid Liquid
MDL No. MFCD00146206 MFCD00146206 MFCD00146252 MFCD00146252 MFCD00147253 MFCD00147253 MFCD00134130 MFCD00163176 MFCD03458582
Appearance Colourless Colourless

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR