Hydrochloric acid ∼37% in water ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

423790025 423795000 423790026 423790251 423795001
423790025.EA 55.7 GBP
423790025. 423795000. 423790026. 423790251. 423795001.
Hydrochloric acid ∼37% in water ACS
Hydrochloric acid
Formula: HCl
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –35 °C
Density: 1.18 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear solution
Color scale ≤10 APHA
Residue after ignition ≤0.0005 %
Titration with NaOH 36.5 to 38.0 %
Bromide (Br) ≤50 ppm
Sulfite (SO3) ≤1 ppm
Sulfate (SO4) ≤1 ppm
Free chlorine ≤1 ppm (as Cl)
Ammonium (NH4) ≤3 ppm
Arsenic (As) ≤0.01 ppm
Iron (Fe) ≤0.2 ppm
Heavy metals (ICP-OES) ≤1 ppm
Extractable organic subst (about 5 ppm) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR