Cobalt ≥99.95% (metals basis), slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

42355.06 42355.14 42355.22
42355.06EA 106 GBP
42355.06 42355.14 42355.22
Cobalt ≥99.95% (metals basis), slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Cobalt

Purity: 99.95% CAS No.: 7440-48-4 Physical Form: ≈0.22g/slug Odour: Odourless MDL No.: MFCD00010935

Formula: Co
Boiling Pt: 2870 °C (1013 hPa)
Melting Pt: 1495 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR