Specpure® Silica, Standard Solution, SiO₂ 1000 µg/ml

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

42225.AE 42225.AP
42225.AEEA 80.5 GBP
42225.AE 42225.AP
Specpure® Silica, Standard Solution, SiO₂ 1000 µg/ml
Standards Anionic Standards Silicate Standards

Molecular Formula: Na₂SiO₃ in H₂O
Physical Form: Liquid

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR