Acetic acid:Trimethylpentane (60:40)% (v/v)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

41895.AEEA 23.6 GBP
41895.AE
Acetic acid:Trimethylpentane (60:40)% (v/v)
Acetic acid:Trimethylpentane

Applications: Testing of peroxide


Molecular Formula: CH3CO2H/(CH3)3CCH2CH(CH3)2 Physical Form: Liquid UN No.: UN2924 MDL No.: MFCD00236632


Applications: Testing of peroxide

Formula: CH₃CO₂H:(CH₃)₃CCH₂CH(CH₃)₂
Storage Temperature: Ambient
MDL Number: MFCD00236632
UN: 2924
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR