Lithiumbis(trimethylsilyl)amide 0.9-1.1 M in hexane

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Lithiumbis(triméthylsilyl)amide, HMDS-Li, N-Lithiohexamethyldisilazane, HMDS lithium salt

Packaged under argon in resealable ChemSeal™ bottles.

41872.22 41872.36
41872.22EA 55.8 GBP
41872.22 41872.36
Lithiumbis(trimethylsilyl)amide 0.9-1.1 M in hexane
Lithium bis(trimethylsilyl)amide

CAS No.: 4039-32-1 Molecular Formula: C6H18LiNSi2 Formula Weight: 167.33 Flash Point: -31°(-24°F) Physical Form: Liquid Density: 0.716 Appearance: Colourless to pale yellow Beilstein Reference: 8136861 UN No.: 2924 MDL No.: MFCD00008261

Formula: C₆H₁₈LiNSi₂
MW: 167.33 g/mol
Density: 0.716
Flash Pt: –31 °C (–24 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008261
CAS Number: 4039-32-1
EINECS: 223-725-6
UN: 2924
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR