Sulfur in residual oil standard solutions, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Sulfur in residual oil standard solutions, Specpure®
Standards Petroleum Industry Standards

Physical Form: Liquid
MDL No.: MFCD00197838

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No. MFCD00197838
Physical form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR