Calcium hexaboride ≥99.5%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

41722.06 41722.14 41722.22
41722.06EA 46.9 GBP
41722.06 41722.14 41722.22
Calcium hexaboride ≥99.5%
Calcium hexaboride

Purity: 99.5% CAS No.: 12007-99-7 Molecular Formula: CaB6 Formula Weight: 104.95 Physical Form: -325 Mesh Powder Density: 2.30 Melting Point: 2235° MDL No.: MFCD00148904

Formula: B₆Ca
Melting Pt: 2235 °C
Density: 2.30
Storage Temperature: Ambient
MDL Number: MFCD00148904
CAS Number: 12007-99-7
EINECS: 234-525-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR