Multi Ion anion IC standard solution in H₂O, Specpure®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

41693.AEEA 231 GBP
41693.AE
Multi Ion anion IC standard solution in H₂O, Specpure®
Standards Anionic Standards Multi-Element Anioinic Standards

Physical Form: Liquid
Odour: Odourless
MDL No.: MFCD00144814

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR