Sulphanilic acid 98.0-102.0% ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

41540.18 41540.30
41540.18EA 27.6 GBP
41540.18 41540.30
Sulphanilic acid 98.0-102.0% ACS
Sulphanilic acid

Purity: 98.0-102.0% CAS No.: 121-57-3 Molecular Formula: C6H7NO3S Formula Weight: 173.19 Physical Form: Powder Melting Point: ca 365° Beilstein Reference: 908765 Merck Reference: 14,8926 MDL No.: MFCD00007886


Reagent for chromatographic detection of histidine

Formula: 4-(H₂N)C₆H₄SO₃H
Boiling Pt: 363 °C (1013 hPa)
Melting Pt: ∼365 °C
Density: 1.485 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007886
CAS Number: 121-57-3
EINECS: 204-482-5
Merck Index: 14,8926
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e032bed891f24ee48419dc3c7237b0293fd3e629&vid=28238ab814bb2b8d6eb5012387d8bf41528469e7&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR