Polyvinyl alcohol, M.W. ∼ 10,000-26,000 86-89, molecular weight_low, hydrolysed

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

41238.14 41238.22 41238.36
41238.14EA 28 GBP
41238.14 41238.22 41238.36
Polyvinyl alcohol, M.W. ∼ 10,000-26,000 86-89, molecular weight_low, hydrolysed
Polyvinyl alcohol

CAS No.: 9002-89-5 Molecular Formula: [-CH2CH(OH)-]n Odour: Odourless MDL No.: MFCD00081922

Formula: [-CH₂CH(OH)-]n
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR