Laboratory grinding mill, MultiDrive basic
Supplier: IKA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
25002640
412-0323EA 4220 GBP
412-0323
Laboratory grinding mill, MultiDrive basic
Mills
Regardless of whether samples are hard, soft or fibrous this MultiDrive crusher can perform a wide range of crushing tasks involving coarse and fine crushing. MultiDrive basic is ideal to mix or grind the samples.
  • Excellent crushing performance is guaranteed by a combination of variable rotational speeds
  • USB interface available for easy actuation and documentation respectively
  • Stainless steel cutter/beater
  • Hard metal cutter/beater for milling chamber
  • Brushless motor
  • In combination with the milling chamber MI 250 MultiDrive basic replaces M 20 mill