Cutting mills, SM 100 and SM 200
Supplier: Retsch GmbH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
20.728.0002 20.727.0002 20.727.0001 20.728.0001 20.727.0009 20.727.0010
412-0289EA 14780 GBP
412-0289 412-0288 510-1373 412-0220 412-0280 412-0281
Cutting mills, SM 100 and SM 200
Mills
The SM 100 and SM 200 are easy to operate, fixed speed, powerful cutting mills for rapid and gentle size reduction of bulky, soft, medium-hard, fibrous, elastic and thermally sensitive materials. Ideal for the batchwise or continuous preliminary size reduction of dry materials such as wood, roots, bones, plants, plastics, feed pellets, spices, drugs, paper and cardboard. Long fibre samples and voluminous materials can be processed without any preliminary size reduction.
  • Rapid and gentle size reduction of dry materials
  • Samples subjected to low thermal
More Product Information