Caffeine, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: 3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 7-Methyltheobromine, 1,3,7-Trimethylxanthine, Methyltheobromine, 1,3,7-Trimethyl-2,6-dioxopurine, 2,6-Dihydroxy-1,3,7-trimethylpurine

MQ100

41019-5G 41019-1G
41019-5GEA 40.6 GBP
41019-5G 41019-1G
Caffeine, Supelco®
Caffeine
Formula: C₈H₁₀N₄O₂
MW: 194.19 g/mol
Melting Pt: 235…239 °C
Density: 1.23 g/cm³ (19 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005758
CAS Number: 58-08-2
EINECS: 200-362-1
Merck Index: 13,01636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01f32f2961a9624593ad2208ea5ec8df149b7831&vid=6e01f5468289365f67995e462a55d79debd3e6b3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR