Bismuth(III) hydroxide, tech.

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

40397.30 40397.A1
40397.30EA 77.7 GBP
40397.30 40397.A1
Bismuth(III) hydroxide, tech.
Bismuth (III) hydroxide

CAS No.: 10361-43-0 Molecular Formula: Bi(OH)3 Formula Weight: 260.00 Physical Form: Amorphous Powder Odour: Odourless Solubility: Insoluble in water. Soluble in acids MDL No.: MFCD00046188


Hydrolysis of ribonucleic acid, absorbent

Formula: H₃BiO₃
Storage Temperature: Ambient
MDL Number: MFCD00046188
CAS Number: 10361-43-0
EINECS: 233-790-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR