Gold 14kt (Au ≥ 58.3%), yellow, wire, Ø 1.63 mm (0.064 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

40329.BS 40329.BU
40329.BSEA 430 GBP
40329.BS 40329.BU
Gold 14kt (Au ≥ 58.3%), yellow, wire, Ø 1.63 mm (0.064 in)
Gold

Molecular Formula: % Concentration CuZnAg Physical Form: ≈28g/m Appearance: Yellow MDL No.: MFCD00003436

Formula: Au
Boiling Pt: 2800 °C (1013 hPa)
Melting Pt: 1063 °C
Density: 19.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9
Merck Index: 12,04526

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR