Erbium ≥99.9% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

40287.DH 40287.DJ 40287.HF
40287.DHEA 273 GBP
40287.DH 40287.DJ 40287.HF
Erbium ≥99.9% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)
Erbium

Purity: 99.9% CAS No.: 7440-52-0 Molecular Formula: Packaged in mylar under argon Physical Form: ≈0.29g/mm, Cast rod, diameter is approximate Odour: Odourless

Formula: Er
Storage Temperature: Ambient
MDL Number: MFCD00010987
CAS Number: 7440-52-0
EINECS: 231-160-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR