Formamide 99,5 GC_ASSAY_METHOD, ultrapure_bioreagent nucleic acids_denaturing prior analysis, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Carbamaldehyde

Nitrogen_filled

4028-00EA 72.9 GBP
4028-00 4028-01
Formamide 99,5 GC_ASSAY_METHOD, ultrapure_bioreagent nucleic acids_denaturing prior analysis, J.T.Baker®
Formamide

For denaturing nucleic acids prior to analysis.


ULTRAPURE BIOREAGENT—A line of high purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography. Where applicable, tests include DNase, RNase, Protease, heavy metals and insoluble matter.

Formula: HCONH₂
MW: 45.04 g/mol
Boiling Pt: 210
Melting Pt: 2
Density: 1.1334 g/cm³ (20 °C)
Flash Pt: 79
Storage Temperature: Refrigerator
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0
Merck Index: 13,04261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use = 99.5 %
Assay (HCONH₂) (by GC) ≥99.5 %
Absorbance of a 1M Solution - 280 nm ≤0.100
Absorbance of a 1M Solution - Color (APHA) ≤10
Absorbance of a 1M Solution - Conductivity, mS/cm (1:10), For Information Only 2.0 - 3.0 °C
Freezing Point 2.0 - 3.0 °C
Trace Impurities - Copper (Cu) ≤0.1 ppm
Trace Impurities - Iron (Fe) ≤0.5 ppm
Trace Impurities - Lead (Pb) ≤0.5 ppm
Trace Impurities - Zinc (Zn) ≤0.5 ppm
Packaged under Nitrogen
Deionization of material is not required.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR