Copper(I) thiocyanate 96

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Copper thiocyanate, Copper (I) rhodanide, Cuprous thiocyanate, Copper rhodanide

40220.18 40220.30
40220.18EA 36.5 GBP
40220.18 40220.30
Copper(I) thiocyanate 96
Copper (I) thiocyanate

Purity: 96% min CAS No.: 1111-67-7 Molecular Formula: CuSCN Formula Weight: 121.63 Physical Form: Powder Density: 2.84 Melting Point: 1084° Solubility: Practically insoluble in water, alcohol. Soluble in NH4OH, ether Merck Reference: 14,2669 UN No.: 3077 MDL No.: MFCD00010980

Formula: CCuNS
MW: 121.63 g/mol
Density: 2.84
Storage Temperature: Ambient
MDL Number: MFCD00010980
CAS Number: 1111-67-7
EINECS: 214-183-1
UN: 3077
ADR: 9 III
Merck Index: 13,02699

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR