Palladium ≥99.98% (metals basis), wire, Ø 1.0 mm

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

40212.BS 40212.BW 40212.CB
40212.BSEA 284 GBP
40212.BS 40212.BW 40212.CB
Palladium ≥99.98% (metals basis), wire, Ø 1.0 mm
Palladium

Purity: 99.98+% CAS No.: 7440-05-3 Physical Form: ≈9.44g/m Odour: Odourless MDL No.: MFCD00011167

Formula: Pd
Boiling Pt: 3167 °C (1013 hPa)
Melting Pt: 1555 °C
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
Merck Index: 12,07121

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR