Cadmium(II) fluoride ≥99.99% (metals basis), Puratronic®

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

40203.06 40203.14 40203.22
40203.06EA 36.8 GBP
40203.06 40203.14 40203.22
Cadmium(II) fluoride ≥99.99% (metals basis), Puratronic®
Cadmium (II) fluoride

Purity: 99.99% CAS No.: 7790-79-6 Molecular Formula: CdF2 Formula Weight: 150.40 Physical Form: Powder Density: 6.64 Melting Point: 1100° Boiling Point: 1758° Merck Reference: 14,1619 UN No.: 2570 MDL No.: MFCD00010919

Formula: CdF₂
Boiling Pt: 1758 °C
Melting Pt: 1100 °C
Density: 6.64
Storage Temperature: Ambient
MDL Number: MFCD00010919
CAS Number: 7790-79-6
EINECS: 232-222-0
UN: 2570
ADR: 6.1,II
Merck Index: 14,1619

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR