7-Aminoactinomycin D (7-AAD)

Supplier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: 2,7-Diamino-N,N'-bis[(6S,9R,10S,13R,18aS)-6,13-diisopropyl-2,5,9-trimethyl-1,4,7,11,14-pentaoxohexadecahydro-1H-pyrrolo[2,1-i][1,4,7,10,13]oxatetraazacyclohexadecin-10-yl]-4,6-dimethyl-3-oxo-3H-phenox azine-1,9-dicarboxamide, Actinomy, 7-Aminoactinomycin D, NSC-239759, NSC 239759, 2,7-diamino-4,6-dimethyl-3-oxo-1-N,9-N-bis[7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]phenoxazine-1,9-dicarboxamide, 7-AAD

40037
40037.EA 214 GBP
40037.
7-Aminoactinomycin D (7-AAD)
7-Aminoactinomycin D (7-AAD)
Formula: C₆₂H₈₇N₁₃O₁₆
MW: 1270.43 g/mol
MDL Number: MFCD00077295
CAS Number: 7240-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR