Cadmium ≥99.9% (metals basis), shot, Ø 5 cm (2 in)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

39731.KT
39731.KTEA 65.6 GBP
39731.KT
Cadmium ≥99.9% (metals basis), shot, Ø 5 cm (2 in)
Cadmium

Purity: 99.9% CAS No.: 7440-43-9 Physical Form: ≈600g/shot MDL No.: MFCD00010914

Formula: Cd
Boiling Pt: 765 °C (1013 hPa)
Melting Pt: 321 °C
Density: 8.642 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010914
CAS Number: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1,II
Merck Index: 12,01649

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR