Gadolinium hexaboride ≥99.5% (metals basis)

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

39225.03 39225.06
39225.03EA 29.4 GBP
39225.03 39225.06
Gadolinium hexaboride ≥99.5% (metals basis)
Gadolinium hexaboride

Purity: 99.5% CAS No.: 12008-06-9 Molecular Formula: GdB6 Formula Weight: 222.12 Physical Form: -325 Mesh Powder Odour: Odourless MDL No.: MFCD00049942

Formula: B₆Gd
Storage Temperature: Ambient
MDL Number: MFCD00049942
CAS Number: 12008-06-9
EINECS: 234-530-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR